Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

565768

Sigma-Aldrich

Poly(ethylene oxide), 4-arm, succinimidyl glutarate terminated

average Mn 10,000 by GPC

Synonym(s):

PEO-4 arm-SG terminated

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12162002

mol wt

average Mn 10,000 by GPC

reaction suitability

reactivity: amine reactive

Mw/Mn

<1.2

polymer architecture

shape: 4-arm
functionality: homofunctional

storage temp.

2-8°C

Application

Useful in biodegradable drug delivery applications.

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service