Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

564567

Sigma-Aldrich

Poly(dimethylsiloxane)-graft-polyacrylates

Mn ~26,000

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352300

form

powder

mol wt

Mn ~26,000

composition

dimethylsiloxane, ~80 wt. %

solubility

H2O: soluble
hydroalcohols: methoxyisopropanol: soluble

Application

Additive for films and coatings for surface modification of
paper, plastics, and metals.

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service