Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

563730

Sigma-Aldrich

4-n-Propylphenylmagnesium bromide solution

0.5 M in THF

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H4CH2CH2CH3MgBr
CAS Number:
Molecular Weight:
223.39
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

reaction suitability

reaction type: Grignard Reaction

concentration

0.5 M in THF

bp

65-67 °C (lit.)

density

0.940 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

CCCc1ccc([Mg]Br)cc1

InChI

1S/C9H11.BrH.Mg/c1-2-6-9-7-4-3-5-8-9;;/h4-5,7-8H,2,6H2,1H3;1H;/q;;+1/p-1

InChI key

HFPDWJMICZDKNO-UHFFFAOYSA-M

Legal Information

Product of Rieke Metals, Inc.
Rieke is a registered trademark of Rieke Metals, Inc.

signalword

Danger

Hazard Classifications

Acute Tox. 4 Oral - Carc. 2 - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1B - STOT SE 3

target_organs

Central nervous system, Respiratory system

supp_hazards

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

1.4 °F - closed cup

flash_point_c

-17 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service