Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

563544

Sigma-Aldrich

3-(Trifluoromethoxy)iodobenzene

96%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H4OCF3I
Molecular Weight:
288.01
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

96%

form

solid

refractive index

n20/D 1.5200 (lit.)

bp

185-186 °C (lit.)

density

1.863 g/mL at 25 °C (lit.)

SMILES string

FC(F)(F)Oc1cccc(I)c1

InChI

1S/C7H4F3IO/c8-7(9,10)12-6-3-1-2-5(11)4-6/h1-4H

Inchi Key

UQZXQSQWKJZHCD-UHFFFAOYSA-N

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

135.0 °F - closed cup

flash_point_c

57.2 °C - closed cup

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service