Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

56350

Sigma-Aldrich

5-Hydroxy-2-pyrazinecarboxylic acid

≥96.0% (T)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H4N2O3
CAS Number:
Molecular Weight:
140.10
Beilstein/REAXYS Number:
775641
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

assay

≥96.0% (T)

form

solid

impurities

≤5.0% water

SMILES string

OC(=O)c1cnc(O)cn1

InChI

1S/C5H4N2O3/c8-4-2-6-3(1-7-4)5(9)10/h1-2H,(H,7,8)(H,9,10)

InChI key

CGQFCIHUUCMACC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Mehmedagic et al.
Journal of chromatography. B, Biomedical sciences and applications, 695(2), 365-372 (1997-08-01)
A new high-performance liquid chromatograhic procedure for simultaneous determination of pyrazinamide (PZA) and its three metabolites 5-hydroxypyrazinamide (5-OH-PZA), pyrazinoic acid (PA), and 5-hydroxypyrazinoic acid (5-OH-PA), in rat urine was developed. 5-OH-PZA and 5-OH-PA standards were obtained by enzymatic synthesis (xanthine
C Lacroix et al.
Journal of chromatography, 422, 217-225 (1987-11-27)
The method reported here for determining pyrazinamide and its metabolites (2-pyrazinoic acid, 5-hydroxypyrazinamide, 5-hydroxypyrazinoic acid and pyrazinuric acid) consists of diluting urine or acid deproteinisation of serum followed by chromatography on a cation-exchange column. The column length and the detection
T Yamamoto et al.
International journal of clinical pharmacology and therapeutics, 33(6), 360-365 (1995-06-01)
We investigated the effect of xylitol on the plasma concentration and the urinary excretion of purine bases, 5-hydroxypyrazinamide and 5-hydroxypyrazinoic acid in subjects who had ingested pyrazinamide (60 mg/kg weight). One liter of 10% xylitol was infused intravenously over 2
C Lacroix et al.
Arzneimittel-Forschung, 40(1), 76-79 (1990-01-01)
The pharmacokinetics of pyrazinamide (Pirilène) and its metabolites are evaluated in ten subjects with hepatic insufficiency, after an oral dose of 19.3 +/- 0.6 mg.kg-1 and the results are compared to those of a group of nine healthy subjects (control
Edgar A Florentini et al.
PloS one, 15(11), e0241600-e0241600 (2020-11-06)
Pyrazinamide (PZA) susceptibility testing in Mycobacterium tuberculosis (Mtb) is a current area of development and PZA-resistant strains are increasingly prevalent. Previous studies have demonstrated that the detection of pyrazinoic acid (POA), the metabolite produced by the deamidation of PZA, is

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service