Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

56193

Sigma-Aldrich

(S)-3-Hydroxy-3-phenylpropionic acid

≥98.0% (sum of enantiomers, GC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H10O3
CAS Number:
Molecular Weight:
166.17
Beilstein/REAXYS Number:
3197773
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:

assay

≥98.0% (sum of enantiomers, GC)

form

solid

optical activity

[α]20/D −22.5±1°, c = 4% in methanol

mp

117-120 °C

storage temp.

2-8°C

SMILES string

O[C@@H](CC(O)=O)c1ccccc1

InChI

1S/C9H10O3/c10-8(6-9(11)12)7-4-2-1-3-5-7/h1-5,8,10H,6H2,(H,11,12)/t8-/m0/s1

InChI key

AYOLELPCNDVZKZ-QMMMGPOBSA-N

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kati Hanhineva et al.
PloS one, 7(6), e39322-e39322 (2012-06-30)
The human gut hosts a microbial community which actively contributes to the host metabolism and has thus remarkable effect on our health. Intestinal microbiota is known to interact remarkably with the dietary constituents entering the colon, causing major metabolic conversions

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service