Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

560545

Sigma-Aldrich

3,5-Dimethoxyphenethylamine

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3O)2C6H3CH2CH2NH2
CAS Number:
Molecular Weight:
181.23
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

98%

drug control

USDEA Schedule I

refractive index

n20/D 1.5100 (lit.)

bp

178-180 °C/19 mmHg (lit.)

density

>1.041 g/mL at 25 °C (lit.)

SMILES string

COc1cc(CCN)cc(OC)c1

InChI

1S/C10H15NO2/c1-12-9-5-8(3-4-11)6-10(7-9)13-2/h5-7H,3-4,11H2,1-2H3

Inchi Key

ZHSFEDDRTVLPHH-UHFFFAOYSA-N

General description

3,5-Dimethoxyphenethylamine is a positional isomer of dimethoxyphenethylamine. Mass spectral studies suggest that on bromination, 3,5-dimethoxyphenethylamine produces 2,6-dibromo product.[1]

Pictograms

Corrosion

signalword

Danger

hcodes

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class

8A - Combustible corrosive hazardous materials

wgk_germany

WGK 3

flash_point_f

230.0 °F - closed cup

flash_point_c

110 °C - closed cup

ppe

Faceshields, Gloves, Goggles, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gas Chromatographic-Mass Spectrometric and High-Performance Liquid Chromatographic Analyses of the Bromination Products of the Regioisomeric Dimethoxyphenethylamines: Differentiation of Nexus from Five Positional Isomers.
DeRuiter JC, et al.
Journal of Chromatographic Science, 36(1), 23-28 (1998)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service