Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

560537

Sigma-Aldrich

4-Ethylphenethylamine

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2C6H4CH2CH2NH2
CAS Number:
Molecular Weight:
149.23
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

97%

refractive index

n20/D 1.5230 (lit.)

bp

240-241 °C (lit.)

bulk density

0.940 g/mL

functional group

amine

SMILES string

CCc1ccc(CCN)cc1

InChI

1S/C10H15N/c1-2-9-3-5-10(6-4-9)7-8-11/h3-6H,2,7-8,11H2,1H3

InChI key

XLJAVPNHXCHBPU-UHFFFAOYSA-N

General description

4-Ethylphenethylamine is present in the essential oil obtained from the stem bark extract of Psidium guajava.

pictograms

Corrosion

signalword

Danger

hcodes

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class

8A - Combustible corrosive hazardous materials

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Faceshields, Gloves, Goggles, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chemical composition, toxicity and antioxidant activities of essential oils of stem bark of Nigerian species of guava (Psidium guajava Linn.).
Fasola TR, et al.
EXCLI Journal , 10, 34-43 (2011)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service