Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

560138

Sigma-Aldrich

Trimethylsilyl acrylate

Synonym(s):

Acrylic acid trimethylsilanyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH2=CHCO2Si(CH3)3
CAS Number:
Molecular Weight:
144.24
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

refractive index

n20/D 1.4090 (lit.)

bp

53 °C/0.2 mmHg (lit.)

density

0.984 g/mL at 25 °C (lit.)

SMILES string

C[Si](C)(C)OC(=O)C=C

InChI

1S/C6H12O2Si/c1-5-6(7)8-9(2,3)4/h5H,1H2,2-4H3

InChI key

OTYBJBJYBGWBHB-UHFFFAOYSA-N

signalword

Danger

Hazard Classifications

Aquatic Chronic 2 - Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

3 - Flammable liquids

wgk_germany

WGK 2

flash_point_f

5.0 °F - closed cup

flash_point_c

-15 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service