Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

559806

Sigma-Aldrich

4-Amino-3-methyl-6-phenyl-1,2,4-triazin-5-one

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H10N4O
CAS Number:
Molecular Weight:
202.21
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

97%

form

solid

mp

167-170 °C (lit.)

SMILES string

CC1=NN=C(c2ccccc2)C(=O)N1N

InChI

1S/C10H10N4O/c1-7-12-13-9(10(15)14(7)11)8-5-3-2-4-6-8/h2-6H,11H2,1H3

Inchi Key

VHCNQEUWZYOAEV-UHFFFAOYSA-N

General description

4-Amino-3-methyl-6-phenyl-1,2,4-triazin-5-one, commonly known as metamitron, is a pesticide having low polarity and is hydrophobic in nature. Its pKa value is 2.9.

Pictograms

Skull and crossbonesEnvironment

signalword

Danger

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 3 Dermal - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Faceshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Adsorption of pesticides carbetamide and metamitron on organohydrotalcite.
Bruna F, et al.
Applied Clay Science, 33(2), 116-124 (2006)
Caprylate intercalated layered double hydroxide as adsorbent of the linuron, 2, 4-DB and metamitron pesticides from aqueous solution.
Pavlovic I, et al.
Applied Clay Science, 80, 76-84 (2013)
Heike Thiel et al.
Pest management science, 66(9), 1011-1017 (2010-08-24)
Resistance to photosystem II inhibitors-triazines (atrazine) and triazinones (metamitron, metribuzin)-in Chenopodium album L. is caused by the serine 264 to glycine mutation in the D1 protein. This mutation has been detected in C. album collections from Belgium with unsatisfactory metamitron
E Mechant et al.
Communications in agricultural and applied biological sciences, 70(3), 501-506 (2006-04-28)
Metamitron is a key herbicide in modern low rate weed control programs in sugar beet. Fat hen (Chenopodium album, CHEAL) is a common, highly competitive, weed in sugar beet and one of the targets of metamitron. Recently, unsatisfactory control of
E Mechant et al.
Communications in agricultural and applied biological sciences, 71(3 Pt A), 725-732 (2007-03-30)
In recent years, in several of the Belgian sugar beet growing regions, farmers have been confronted with unsatisfactory control of fat hen (Chenopodium album L.). Greenhouse bioassays conducted on reference C. album populations and on "suspected" populations from sugar beet

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service