Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

558907

Sigma-Aldrich

Aluminum tri-sec-butoxide solution

1.0 M in methylene chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Al[OCH(CH3)C2H5]3
Molecular Weight:
246.32
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

form

solution

reaction suitability

core: aluminum

concentration

1.0 M in methylene chloride

bp

40 °C

density

1.237 g/mL at 25 °C

SMILES string

CCC(C)O[Al](OC(C)CC)OC(C)CC

InChI

1S/3C4H9O.Al/c3*1-3-4(2)5;/h3*4H,3H2,1-2H3;/q3*-1;+3

InChI key

WOZZOSDBXABUFO-UHFFFAOYSA-N

pictograms

Health hazardExclamation mark

signalword

Warning

Hazard Classifications

Carc. 2 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Central nervous system

wgk_germany

WGK 3

ppe

Eyeshields, Faceshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kwan Wee Tan et al.
ACS nano, 8(5), 4730-4739 (2014-04-02)
Structure control in solution-processed hybrid perovskites is crucial to design and fabricate highly efficient solar cells. Here, we utilize in situ grazing incidence wide-angle X-ray scattering and scanning electron microscopy to investigate the structural evolution and film morphologies of methylammonium

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service