About This Item
assay
98%
reaction suitability
reagent type: catalyst
core: antimony
mp
184 °C (dec.) (lit.)
SMILES string
C[Sb](C)(C)(Br)Br
InChI
1S/3CH3.2BrH.Sb/h3*1H3;2*1H;/q;;;;;+2/p-2
InChI key
HGGPVCKBWYYOBU-UHFFFAOYSA-L
signalword
Warning
hcodes
Hazard Classifications
Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 2
Storage Class
8A - Combustible corrosive hazardous materials
wgk_germany
WGK 2
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service