Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

556882

Sigma-Aldrich

1-(4-Chlorophenyl)-3-methyl-2-pyrazolin-5-one

97%

Synonym(s):

1-(4-Chlorophenyl)-3-methyl-5-pyrazolone, 2-(4-Chlorophenyl)-2,4-dihydro-5-methyl-3H-pyrazol-3-one, 3-Methyl-1-(4-chlorophenyl)-5-pyrazolone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H9ClN2O
CAS Number:
Molecular Weight:
208.64
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

assay

97%

form

solid

mp

167-171 °C (lit.)

SMILES string

CC1=NN(C(=O)C1)c2ccc(Cl)cc2

InChI

1S/C10H9ClN2O/c1-7-6-10(14)13(12-7)9-4-2-8(11)3-5-9/h2-5H,6H2,1H3

InChI key

WHIXQFSPEDIMGL-UHFFFAOYSA-N

General description

1-(4-Chlorophenyl)-3-methyl-2-pyrazolin-5-one is an analog of edaravone, a radioprotector. The similarity in the absorption spectra of 1-(4-chlorophenyl)-3-methyl-2-pyrazolin-5-one radical edaravone radical indicates that the phenyl group is involved in the reaction with the OH. radicals. Absorption spectrum of 1-(4-chlorophenyl)-3-methyl-2-pyrazolin-5-one radical oxidized by N3 shows an absorption peak at 360nm.[1]

Application

1-(4-Chlorophenyl)-3-methyl-2-pyrazolin-5-one may be used to synthesize 1-(2-chlorophenyl)-3-methylchromeno[2,3-c]pyrazol-4(1H)-one.[2]

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kuniki Hata et al.
Journal of radiation research, 52(1), 15-23 (2010-12-09)
A comparative study using the pulse radiolysis technique was carried out to investigate transient absorption spectra and rate constants for the reactions of (•)OH and N(3)(•) with edaravone (3-methyl-1-phenyl-2-pyrazolin-5-one) and its four analogue compounds, 1,3-dimethyl-2-pyrazolin-5-one, 3-methyl-1-(pyridin-2-yl)-2-pyrazolin-5-one, 1-phenyl-3-trifluoromethyl-2-pyrazolin-5-one and 1-(4-chlorophenyl)-3-methyl-2-pyrazolin-5-one. The
(2-Chlorophenyl)-3-methylchromeno [2, 3-c] pyrazol-4 (1H)-one.
Batezila G and Holzer W.
Molbank, 2010(1), M661-M661 (2010)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service