Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

552925

Sigma-Aldrich

2-Methylthiophenothiazine

97%

Synonym(s):

2-Methylsulfanyl-10H-phenothiazine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H11NS2
CAS Number:
Molecular Weight:
245.36
EC Number:
MDL number:
UNSPSC Code:
12352303
PubChem Substance ID:
NACRES:
NA.23

assay

97%

mp

140-144 °C (lit.)

SMILES string

CSc1ccc2Sc3ccccc3Nc2c1

InChI

1S/C13H11NS2/c1-15-9-6-7-13-11(8-9)14-10-4-2-3-5-12(10)16-13/h2-8,14H,1H3

InChI key

OBVKBOLDEFIQDP-UHFFFAOYSA-N

signalword

Warning

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Skin Sens. 1 - STOT RE 2 Oral

target_organs

Blood

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service