Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

552321

Sigma-Aldrich

2-[(Trimethylsilyl)ethynyl]thiophene

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H12SSi
CAS Number:
Molecular Weight:
180.34
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

97%

refractive index

n20/D 1.549 (lit.)

bp

95-103 °C/10 mmHg (lit.)

density

1.09 g/mL at 25 °C (lit.)

SMILES string

C[Si](C)(C)C#Cc1cccs1

InChI

1S/C9H12SSi/c1-11(2,3)8-6-9-5-4-7-10-9/h4-5,7H,1-3H3

InChI key

OQUBLKNISPLGJP-UHFFFAOYSA-N

pictograms

Flame

signalword

Warning

hcodes

Hazard Classifications

Flam. Liq. 3

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

78.1 °F - closed cup

flash_point_c

25.6 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis of silicon-containing unsaturated polymers by hydrosilylation reactions. Photophysical studies.
Wang F, et al.
Macromolecules, 36(22), 8225-8230 (2003)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service