Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

551716

Sigma-Aldrich

Thiophosphoryl-PMMH-24 dendrimer, generation 3.5

Synonym(s):

Thiophosphoryl-phenoxymethyl(methylhydrazono) dendrimer, generation 3.5

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
S=P(Cl)3:(G=3.5);dendri Thiophosphoryl-PMMH[O(C6H4)CHO]24
CAS Number:
Molecular Weight:
7404.98
MDL number:
UNSPSC Code:
12162002

description

aldehyde surface groups

form

powder

mol wt

generation 3.5

feature

core type Phosphorous dendrimers with thiophosphoryl chloride core

no. Surface Groups

24

mp

181-188 °C (lit.)

Physical form

Contains 24 surface aldehyde groups.

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service