Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

550736

Sigma-Aldrich

2-Isopropylanisole

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHC6H4OCH3
CAS Number:
Molecular Weight:
150.22
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

98%

refractive index

n20/D 1.508 (lit.)

bp

198-200 °C (lit.)

density

0.936 g/mL at 25 °C (lit.)

SMILES string

COc1ccccc1C(C)C

InChI

1S/C10H14O/c1-8(2)9-6-4-5-7-10(9)11-3/h4-8H,1-3H3

InChI key

NNZRVXTXKISCGS-UHFFFAOYSA-N

General description

2-Isopropylanisole can be prepared from 2-isopropylphenol via O-methylation using dimethyl sulfate.[1]

Application

2-Isopropylanisole may be used in the preparation of 4-bromo-2-isopropylanisole via bromination using lithium bromide/ammonium cerium(IV) nitrate system.[1] It may also be used as a starting material in the multi-synthesis of (±)-shonanyl methyl ether.[2]

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

187.0 °F - closed cup

flash_point_c

86.1 °C - closed cup

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Total synthesis of (?)-shonanyl methyl ether and (?)-ferruginyl methyl ether.
Ghosal M, et al.
Tetrahedron Letters, 30(26), 3469-3470 (1989)
Total Syntheses of Multicaulins via Oxidative Photocyclization of Stilbenes.
Sec?inti H, et al.
Journal of Natural Products, 77(9), 2134-2137 (2014)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service