Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

549681

Sigma-Aldrich

4-Acetamido-2-methylbenzonitrile

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CONHC6H4CN
CAS Number:
Molecular Weight:
174.20
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

assay

97%

mp

161-164 °C (lit.)

SMILES string

CC(=O)Nc1ccc(C#N)c(C)c1

InChI

1S/C10H10N2O/c1-7-5-10(12-8(2)13)4-3-9(7)6-11/h3-5H,1-2H3,(H,12,13)

InChI key

SRCREJLHAPEANJ-UHFFFAOYSA-N

General description

4-Acetamido-2-methylbenzonitrile [N-(4-cyano-3-methylphenyl)acetamide] can be prepared by the solvent-free N-acetylation of 4-amino-2-methylbenzonitrile in the presence of a solid acid catalyst.

Application

4-Acetamido-2-methylnitrobenzene may be used in the preparation of 2-nitro-4-aminotoluene by acid hydrolysis.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Modified mesoporous aluminophosphate as an efficient solid acid catalyst for the synthesis of novel O-and N-acetylated compounds: solvent free condition.
Shyamprasad K, et al.
Journal of Porous Materials, 23(4), 1095-1105 (2016)
The Monoacetylation of 2, 4-Diamino-6-nitrotoluene.
1 Foster RJ, et al.
Journal of the American Chemical Society, 72(9), 3959-3960 (1950)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service