549347
4-Carboxybenzenesulfonamide, polymer-bound
100-200 mesh, extent of labeling: 0.7-1.3 mmol/g loading, 1 % cross-linked
Synonym(s):
4-Sulfamylbenzoyl resin, Kenner’s "Safety-Catch"resin
About This Item
Recommended Products
crosslinking
1 % cross-linked
reaction suitability
reaction type: Fmoc solid-phase peptide synthesis
extent of labeling
0.7-1.3 mmol/g loading
particle size
100-200 mesh
functional group
sulfonamide
SMILES string
C=Cc1ccccc1.C=Cc2ccc(C=C)cc2.NS(=O)(=O)c3ccc(cc3)C(=O)NCc4ccccc4
Looking for similar products? Visit Product Comparison Guide
Storage Class
11 - Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Related Content
Ellman group developed electron-rich phosphine ligands for C-H functionalization and tert-Butanesulfinamide for asymmetric amine synthesis.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service