All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C14H9ClN2O
CAS Number:
Molecular Weight:
256.69
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
assay
97%
mp
270-274 °C (lit.)
SMILES string
Clc1ccc(cc1)C2=NNC(=O)c3ccccc23
InChI
1S/C14H9ClN2O/c15-10-7-5-9(6-8-10)13-11-3-1-2-4-12(11)14(18)17-16-13/h1-8H,(H,17,18)
InChI key
ZHLKSRYOYNILEG-UHFFFAOYSA-N
General description
4-(4-Chlorophenyl)-1-(2H)-phthalazinone is a 4-aryl-2(1H)-phthalazinone derivative that can be prepared by refluxing a mixture of 2-(4-chlorobenzoyl)-benzoic acid and hydrazine hydrate in ethanol.
Application
4-(4-Chlorophenyl)-1-(2H)-phthalazinone may be used as a starting material in the synthesis of 1-(N,N-diphenyl-amino)-4-(4-chlorophenyl)-phthalazine, which forms a tris-cyclometalated iridium(III) complex that can act as a dopant in organic light-emitting diodes (OLEDs).
Storage Class
13 - Non Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Synthesis of a novel tris-cyclometalated iridium (III) complex containing triarylamine unit and its application in OLEDs.
Fang Y, et al.
Inorgorganica Chimica Acta, 362(14), 4985-4990 (2009)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service