Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

547778

Sigma-Aldrich

Tris[N,N-bis(trimethylsilyl)amide]lanthanum(III)

98%

Synonym(s):

Lanthanum bis(trimethylsilyl)amide, Lanthanum tris(bis(trimethylsilyl)amide)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
La(N(Si(CH3)3)2)3
CAS Number:
Molecular Weight:
620.06
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.22

assay

98%

form

solid

reaction suitability

core: lanthanum
reagent type: catalyst

mp

149-153 °C (lit.)

SMILES string

C[Si](C)(C)N([La](N([Si](C)(C)C)[Si](C)(C)C)N([Si](C)(C)C)[Si](C)(C)C)[Si](C)(C)C

InChI

1S/3C6H18NSi2.La/c3*1-8(2,3)7-9(4,5)6;/h3*1-6H3;/q3*-1;+3

InChI key

ZDYNTRMQDURVDM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

pictograms

FlameCorrosion

signalword

Danger

Hazard Classifications

Flam. Sol. 1 - Skin Corr. 1B - Water-react 2

supp_hazards

Storage Class

4.3 - Hazardous materials which set free flammable gases upon contact with water

wgk_germany

WGK 3

flash_point_f

36.0 °F - closed cup

flash_point_c

2.2 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type P3 (EN 143) respirator cartridges


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xiaoyan Tang et al.
Nature communications, 9(1), 2345-2345 (2018-06-13)
Bacterial poly(3-hydroxybutyrate) (P3HB) is a perfectly isotactic, crystalline material possessing properties suitable for substituting petroleum plastics, but high costs and low volumes of its production are impractical for commodity applications. The chemical synthesis of P3HB via ring-opening polymerization (ROP) of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service