Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

547697

Supelco

Poly(4-vinylpyridine), cross-linked

Reillex® 425 ion-exchange resin, 25 % cross-linked with divinylbenzene

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
23151816
NACRES:
SB.52

form

beads

crosslinking

25 % cross-linked with divinylbenzene

manufacturer/tradename

Reillex® 425 ion-exchange resin

technique(s)

LPLC: suitable
ion chromatography: suitable

surface area

750 m2/g

matrix

poly(4-vinylpyridine)-divinylbenzene

matrix active group

polymer

particle size

18-50 mesh

pore size

60 Å mean pore size

separation technique

reversed phase

InChI

1S/C10H10.C7H7N/c1-3-9-7-5-6-8-10(9)4-2;1-2-7-3-5-8-6-4-7/h3-8H,1-2H2;2-6H,1H2

InChI key

HUEZUDXJCGHIHG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Poly(4-vinylpyridine) (PVP) is an effective matrix for the incorporation of transition-metal ions into polymer electrodes. Cross-linking of PVP was done by heating mixture of linear polymer, triallyl-substituted cross-linking agent and radical initiator.[1] PVP can be cross-linked by necleophilic reaction.[2]

application

Poly(4-vinylpyridine) may be used as packing material for column chromatography.

Legal Information

Reillex is a registered trademark of Vertellus Specialties, Inc.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Electrodeposition and characterization of platinum microparticles in poly (4-vinylpyridine) film electrodes.
Bartak, Duane E., et al.
Analytical Chemistry, 58, 2756-2761 (1986)
Electrodeposition of palladium, iridium, ruthenium, and platinum in poly (4-vinylpyridine) films for electrocatalysis.
Kost, Kent M., et al.
Analytical Chemistry, 62, 151-157 (1990)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service