Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

546615

Sigma-Aldrich

Poly[2-(2′,5′-bis(2″-ethylhexyloxy)phenyl)-1,4-phenylenevinylene]

Synonym(s):

BEHP-PPV

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352103
NACRES:
NA.23

form

solid

solubility

THF: soluble
chloroform: soluble
xylene: soluble

fluorescence

λex 400 nm; λem 476 nm in toluene

General description

Light-emitting polymer. More stable to photooxidation than other substituted PPV derivatives.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Johansson, D. et al.
Macromolecules, 33, 2525-2525 (2000)

Articles

LEPs enable a wide range of important applications including sensors, flexible LED displays and lighting devices, optical pump lasers, and potentially polymer diode lasers.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service