Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

546518

Sigma-Aldrich

Poly[2,5-bis(3′,7′-dimethyloctyloxy)-1,4-phenylenevinylene]

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

solubility

toluene, xylene, THF, chloroform, chlorobenzene, cyclohexanone: soluble

fluorescence

λex 480 nm; λem 548 nm in toluene

SMILES string

CC(C)CCCC(C)CCOc1ccc(OCCC(C)CCCC(C)C)c(C=C)c1

Application

Conducting polymer. Processability and film-forming properties found to be superior to MEH-PPV.[1]

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Advanced Materials, 10, 1340-1340 (1998)

Articles

LEPs enable a wide range of important applications including sensors, flexible LED displays and lighting devices, optical pump lasers, and potentially polymer diode lasers.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service