Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

545880

Sigma-Aldrich

Dimethyl sulfoxide-d6

"100%", 99.96 atom % D, contains 0.03 % (v/v) TMS

Synonym(s):

(Methyl sulfoxide)-d6, DMSO-d6, Hexadeuterodimethyl sulfoxide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CD3)2SO
CAS Number:
Molecular Weight:
84.17
Beilstein/REAXYS Number:
1237248
EC Number:
MDL number:
UNSPSC Code:
12191502
PubChem Substance ID:

vapor pressure

0.42 mmHg ( 20 °C)

isotopic purity

99.96 atom % D

assay

≥99.00%

form

liquid

autoignition temp.

573 °F

contains

0.03 % (v/v) TMS

expl. lim.

42 %

packaging

pk of 10 × 0.75 mL

impurities

≤0.250% water
water

refractive index

n20/D 1.476 (lit.)

bp

189 °C (lit.)

mp

20.2 °C (lit.)

density

1.190 g/mL at 25 °C (lit.)

mass shift

M+6

SMILES string

[2H]C([2H])([2H])S(=O)C([2H])([2H])[2H]

InChI

1S/C2H6OS/c1-4(2)3/h1-2H3/i1D3,2D3

InChI key

IAZDPXIOMUYVGZ-WFGJKAKNSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class

10 - Combustible liquids

wgk_germany

WGK 2

flash_point_f

190.4 °F

flash_point_c

88 °C

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service