Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

545783

Sigma-Aldrich

3-(3-Furylmethylthio)-2-methylpyrazine

97%

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
$273.00

$273.00


Check Cart for Availability

Request a Bulk Order
Notify Me

Get notified when this item is ready to ship via email.


Select a Size

Change View
25 G
$273.00

About This Item

Empirical Formula (Hill Notation):
C10H10N2OS
Molecular Weight:
206.26
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$273.00


Check Cart for Availability

Request a Bulk Order
Notify Me

Get notified when this item is ready to ship via email.

assay

97%

refractive index

n20/D 1.599 (lit.)

bp

289-299 °C (lit.)

density

1.201 g/mL at 25 °C (lit.)

functional group

thioether

SMILES string

Cc1nccnc1SCc2ccoc2

InChI

1S/C10H10N2OS/c1-8-10(12-4-3-11-8)14-7-9-2-5-13-6-9/h2-6H,7H2,1H3

Inchi Key

JFHOJNODOBNELT-UHFFFAOYSA-N

General description

3-(3-Furylmethylthio)-2-methylpyrazine, also known as 2-(furfurylthio)-3-methylpyrazine, is a 3-substituted 2-alkylpyrazine derivative.

Pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

230.0 °F - closed cup

flash_point_c

110 °C - closed cup

ppe

Eyeshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service