Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

545740

Sigma-Aldrich

(R)-(−)-2-Fluoro-α-methyl-4-biphenylacetic acid

97%

Synonym(s):

(R)-2-Flurbiprofen, Tarenflurbil

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5C6H3(F)CH(CH3)CO2H
CAS Number:
Molecular Weight:
244.26
EC Number:
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.22

assay

97%

mp

110-113 °C (lit.)

SMILES string

C[C@@H](C(O)=O)c1ccc(c(F)c1)-c2ccccc2

InChI

1S/C15H13FO2/c1-10(15(17)18)12-7-8-13(14(16)9-12)11-5-3-2-4-6-11/h2-10H,1H3,(H,17,18)/t10-/m1/s1

InChI key

SYTBZMRGLBWNTM-SNVBAGLBSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

gamma secretase inhibitor; amyloid Abeta 42 lowering agent

pictograms

Skull and crossbonesHealth hazard

signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Oral - Repr. 2 - Skin Sens. 1B

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Faceshields, Gloves, type P2 (EN 143) respirator cartridges


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Natasja de Bruin et al.
Chirality, 30(5), 632-641 (2018-02-07)
Flurbiprofen (F) is a nonsteroidal anti-inflammatory drug (NSAID) used therapeutically as the racemate of (R)-enantiomer and (S)-enantiomer. The inversion of RF to SF and vice versa was investigated in C57Bl/6 and SJL mice and Dark Agouti and Lewis rats. The
Christensen, D.D.
The Journal of Clinical Psychiatry, 9, 32-32 (2007)
Thomas L Kukar et al.
Nature, 453(7197), 925-929 (2008-06-13)
Selective lowering of Abeta42 levels (the 42-residue isoform of the amyloid-beta peptide) with small-molecule gamma-secretase modulators (GSMs), such as some non-steroidal anti-inflammatory drugs, is a promising therapeutic approach for Alzheimer's disease. To identify the target of these agents we developed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service