Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

545171

Sigma-Aldrich

N,N-Di-N-hexylaniline

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6H13)2NC6H5
CAS Number:
Molecular Weight:
261.45
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

assay

97%

refractive index

n20/D 1.507 (lit.)

bp

165 °C/2 mmHg (lit.)

density

0.89 g/mL at 25 °C (lit.)

SMILES string

CCCCCCN(CCCCCC)c1ccccc1

InChI

1S/C18H31N/c1-3-5-7-12-16-19(17-13-8-6-4-2)18-14-10-9-11-15-18/h9-11,14-15H,3-8,12-13,16-17H2,1-2H3

InChI key

DGYRVXQIGUEFFK-UHFFFAOYSA-N

General description

N,N-Di-N-hexylaniline is N,N-dialkylaniline. It can be prepared by refluxing a mixture of aniline, iodohexane and potassium carbonate in ethanol.[1]

Application

N,N-Di-N-hexylaniline (N,N-dihexylaniline) may be used to synthesize:
  • 4-formyl-N,N-dihexylaniline[1]
  • N,N′-bis[(4-(N,N-dihexylamino)benzylidene]diaminofumaronitrile, a bisazomethine dye[2]
  • 4-(dihexylamino)benzaldehyde[3]

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mutsuo Nuriya et al.
Nature communications, 7, 11557-11557 (2016-05-10)
Second harmonic generation (SHG) imaging can be used to visualize unique biological phenomena, but currently available dyes limit its application owing to the strong fluorescent signals that they generate together with SHG. Here we report the first non-fluorescent and membrane
Star?Shaped Push?Pull Compounds Having 1, 3, 5?Triazine Cores.
Meier H, et al.
European Journal of Organic Chemistry, 2006(11), 2609-2617 (2006)
Design and synthesis of push-pull chromophores for second-order nonlinear optics derived from rigidified thiophene-based ?-conjugating spacers.
Raimundo JM, et al.
Journal of Oral Surgery (American Dental Association : 1965), 67(1), 205-218 (2002)
Alkyl substituent effects on J-or H-aggregate formation of bisazomethine dyes.
Kinashi K, et al.
Dyes and Pigments, 92(1), 783-788 (2012)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service