Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

544337

Sigma-Aldrich

5-Norbornen-2-yl(ethyl)chlorodimethylsilane, mixture of endo and exo

95%

Synonym(s):

2-(5-Norbornenyl)ethyldimethylchlorosilane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H19ClSi
CAS Number:
Molecular Weight:
214.81
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.23

assay

95%

refractive index

n20/D 1.48 (lit.)

bp

231-232 °C (lit.)

density

0.991 g/mL at 25 °C (lit.)

SMILES string

C[Si](C)(Cl)CCC1C[C@H]2C[C@@H]1C=C2

InChI

1S/C11H19ClSi/c1-13(2,12)6-5-11-8-9-3-4-10(11)7-9/h3-4,9-11H,5-8H2,1-2H3/t9-,10+,11?/m1/s1

InChI key

UFEKKYNQUHJSEC-JKIOLJMWSA-N

pictograms

Corrosion

signalword

Danger

hcodes

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class

8A - Combustible corrosive hazardous materials

wgk_germany

WGK 3

flash_point_f

219.9 °F - closed cup

flash_point_c

104.4 °C - closed cup

ppe

Faceshields, Gloves, Goggles, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service