Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

544299

Sigma-Aldrich

4-Bromo-3,5-dimethylisoxazole

95%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H6BrNO
CAS Number:
Molecular Weight:
176.01
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

95%

refractive index

n20/D 1.486 (lit.)

bp

176 °C (lit.)

density

1.478 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

Cc1noc(C)c1Br

InChI

1S/C5H6BrNO/c1-3-5(6)4(2)8-7-3/h1-2H3

InChI key

GYHZPSUAMYIFQD-UHFFFAOYSA-N

General description

4-Bromo-3,5-dimethylisoxazole is also known as 3,5-dimethyl-4-bromoisoxazole.

Application

4-Bromo-3,5-dimethylisoxazole may be used in the preparation of (tert-butyldimethylsilyl)-(3,5-dimethylisoxazol-4-yl)phenylamine[1] and 3,3′,5,5′-tetramethyl-4,4′-diisoxazole.[21}

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Preparation of Heterocyclic Amines via a Copper (I)-Mediated Oxidative Cross-Coupling of Organozinc Reagents with Lithium Amides.
Dunst C, et al.
Synthesis, 2010(13), 2313-2318 (2010)
The Chemistry of Heterocyclic Compounds, 333-333 (2009)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service