Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

542776

Sigma-Aldrich

Ethyl (2R)-(+)-2-[4-(6-chlorobenzoxazol-2-yloxy)phenoxy]propionate

95%

Synonym(s):

Fenoxaprop-P-ethyl

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H16ClNO5
CAS Number:
Molecular Weight:
361.78
Beilstein/REAXYS Number:
8158010
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:

assay

95%

optical activity

[α]20/D +29°, c = 1 in chloroform

mp

79-84 °C (lit.)

SMILES string

[H][C@](C)(Oc1ccc(Oc2nc3ccc(Cl)cc3o2)cc1)C(=O)OCC

InChI

1S/C18H16ClNO5/c1-3-22-17(21)11(2)23-13-5-7-14(8-6-13)24-18-20-15-9-4-12(19)10-16(15)25-18/h4-11H,3H2,1-2H3/t11-/m1/s1

Inchi Key

PQKBPHSEKWERTG-LLVKDONJSA-N

signalword

Warning

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Skin Sens. 1 - STOT RE 2

Storage Class

11 - Combustible Solids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Faceshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ali Es-haghi et al.
Journal of separation science, 38(13), 2305-2311 (2015-04-24)
In this study, organic aerogels were synthesized by the sol-gel polycondensation of mixed cresol with formaldehyde in a slightly basic aqueous solution. Carbon aerogels and xerogels are generated by pyrolysis of organic aerogels. The novel sol-gel-based micro-solid-phase extraction sorbent, resorcinol-formaldehyde
Caroline Rombaldi et al.
Analytical and bioanalytical chemistry, 407(16), 4805-4814 (2015-05-01)
The use of golden mussel shells as a solid support in vortex-assisted matrix solid-phase dispersion (MSPD) was evaluated for the first time for extraction of residues of 11 pesticides and nine pharmaceutical and personal care products from mussel tissue samples.
Sergiane Souza Caldas et al.
Talanta, 146, 676-688 (2015-12-24)
A rapid and efficient sample pretreatment using solvent-based de-emulsification dispersive liquid-liquid microextraction (SD-DLLME) coupled with liquid chromatography-tandem mass spectrometry (LC-MS/MS) was studied for the extraction of 58 pharmaceuticals and personal care products (PPCPs) and pesticides from water samples. Type and
Habib Bagheri et al.
Analytica chimica acta, 886, 56-65 (2015-09-01)
A nanocomposite of silica-polydiphenylamine doped with silver nanoparticles (Ag-SiO2-PDPA) was successfully synthesized by the sol-gel process. For its preparation, PDPA was mixed with butanethiol capped Ag nanoparticles (NPs) and added to the silica sol solution. The Ag NPs were stabilized

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service