Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

541990

Sigma-Aldrich

2′-Chloro-2-hydroxy-5-methylbenzophenone

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClCH6H4COC6H3(OH)CH3
CAS Number:
Molecular Weight:
246.69
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

assay

98%

mp

75-78 °C (lit.)

SMILES string

Cc1ccc(O)c(c1)C(=O)c2ccccc2Cl

InChI

1S/C14H11ClO2/c1-9-6-7-13(16)11(8-9)14(17)10-4-2-3-5-12(10)15/h2-8,16H,1H3

InChI key

BXAARESMPHIJGG-UHFFFAOYSA-N

Application

2′-Chloro-2-hydroxy-5-methylbenzophenone (2-Hydroxy-5-methyl-2′-chlorobenzophenone) may be used in the synthesis of 2-methylxanthene.[1]

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chloro Substituted Diphenylmethanes, Phenyl Benzyl Ethers and Benzophenones Prepared from Ortho-or Para-cresol1, 2.
Huston RC and Robinson KR.
Journal of the American Chemical Society, 73(6), 2483-2486 (1951)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service