Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

541117

Sigma-Aldrich

Triamminemolybdenum(0) tricarbonyl

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(NH3)3Mo(CO)3
CAS Number:
Molecular Weight:
231.06
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

reaction suitability

core: molybdenum
reagent type: catalyst

mp

220-225 °C (lit.)

SMILES string

N.N.N.[Mo].[C-]#[O+].[C-]#[O+].[C-]#[O+]

InChI

1S/3CO.Mo.3H3N/c3*1-2;;;;/h;;;;3*1H3

InChI key

MXBJZAUTHOSABH-UHFFFAOYSA-N

pictograms

Flame

signalword

Danger

hcodes

Hazard Classifications

Water-react 2

supp_hazards

Storage Class

4.3 - Hazardous materials which set free flammable gases upon contact with water

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type P3 (EN 143) respirator cartridges


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service