All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C11H8O3
CAS Number:
Molecular Weight:
188.18
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
Recommended Products
assay
97%
mp
109-112 °C (lit.)
SMILES string
CC(=O)C1C(=O)c2ccccc2C1=O
InChI
1S/C11H8O3/c1-6(12)9-10(13)7-4-2-3-5-8(7)11(9)14/h2-5,9H,1H3
InChI key
SGLKFWMIZOJHCL-UHFFFAOYSA-N
General description
2-Acetyl-1,3-indanedione (AID) is a cyclic β-diketone. It readily forms coordination complexes with lanthanide ions, which display strong luminescence upon their irradiation with ultraviolet light. AID can be prepared from 4-nitrophthalic anhydride.
Storage Class
13 - Non Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Anne Schneider et al.
Journal of neurochemistry, 119(3), 447-459 (2011-08-16)
Coffin-Lowry syndrome is a syndromic form of mental retardation caused by mutations of the Rps6ka3 gene encoding ribosomal s6 kinase (RSK)2. RSK2 belongs to a family containing four members in mammals: RSK1-4. RSKs are serine/threonine kinases and cytosolic substrates of
Europium luminescence in its complexes with 2-acetyl-and 2-benzoyl-1, 3-indanediones.
Vitkun RA, et al.
Doklady Chemistry, 260 (1982)
Efficient Synthesis of 7-Amino-3-hydroxyindan-1-one.
Rajur R, et al.
Synthetic Communications, 39(4), 626-635 (2009)
Christie Buonpane et al.
American journal of physiology. Gastrointestinal and liver physiology, 318(4), G781-G792 (2020-02-25)
Necrotizing enterocolitis (NEC) is a devastating gastrointestinal disease of newborns. Although incompletely understood, NEC is associated with intestinal barrier dysfunction. E-cadherin, an adherens junction, is a protein complex integral in maintaining normal barrier homeostasis. Rho-associated protein kinase-1 (ROCK1) is a
Noriko Oshiro et al.
The Journal of biological chemistry, 289(5), 2658-2674 (2013-12-18)
Activation of mammalian target of rapamycin complex 1 (mTORC1) by amino acids is mediated in part by the Rag GTPases, which bind the raptor subunit of mTORC1 in an amino acid-stimulated manner and promote mTORC1 interaction with Rheb-GTP, the immediate
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service