Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

538345

Sigma-Aldrich

Antimony(III) methoxide

Synonym(s):

Trimethoxyantimony, Trimethoxystibine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Sb(OCH3)3
Molecular Weight:
214.86
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

form

powder

reaction suitability

core: antimony
reagent type: catalyst

mp

123-127 °C (lit.)

SMILES string

CO[Sb](OC)OC

InChI

1S/3CH3O.Sb/c3*1-2;/h3*1H3;/q3*-1;+3

InChI key

KEUYHGXCOWNTEJ-UHFFFAOYSA-N

pictograms

Exclamation markEnvironment

signalword

Warning

pcodes

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 2

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service