Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

536873

Sigma-Aldrich

PAMAM-50% C12 dendrimer, ethylenediamine core, generation 3 solution

20 wt. % in methanol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C686H1376N122O92
CAS Number:
Molecular Weight:
12807.03
MDL number:
UNSPSC Code:
12162002

description

50% amino and 50% [N-(2-hydroxydodecyl)] surface groups

form

liquid

mol wt

generation 3

feature

core type ethylenediamine core (2-carbon core)

concentration

20 wt. % in methanol

no. Surface Groups

32

refractive index

n20/D 1.3615

density

0.8 g/mL at 25 °C

storage temp.

2-8°C

Physical form

Contains ca. 16 primary amino groups and 16 [N-(2-hydroxydodecyl)] surface groups.

Legal Information

Manufactured by Dendritech®, Inc.
Dendritech is a registered trademark of Dendritech, Inc.

signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

target_organs

Eyes

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

51.8 °F

flash_point_c

11 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service