Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

536865

Sigma-Aldrich

PAMAM dendrimer solution

25% C₁₂, generation 3, 20 wt. % in methanol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C494H992N122O76
CAS Number:
Molecular Weight:
9857.93
MDL number:
UNSPSC Code:
12162002
NACRES:
NA.23

Product Name

PAMAM-25% C12 dendrimer, generation 3 solution, 20 wt. % in methanol

description

75% amino and 25% [N-(2-hydroxydodecyl)] surface groups

form

liquid

mol wt

generation 3

feature

core type ethylenediamine core (2-carbon core)

concentration

20 wt. % in methanol

no. Surface Groups

32

refractive index

n20/D 1.363

density

0.8 g/mL at 25 °C

storage temp.

2-8°C

Physical form

Contains ca. 24 primary amino surface groups and 8 [N-(2-hydroxydodecyl)] surface groups

Legal Information

Manufactured by Dendritech®, Inc.
Dendritech is a registered trademark of Dendritech, Inc.

signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

target_organs

Eyes

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

51.8 °F

flash_point_c

11 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service