Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

536806

Sigma-Aldrich

PAMAM dendrimer, ethylenediamine core, generation 7.5 solution

5 wt. % in methanol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[NH2(CH2)2NH2]:(G=7.5);dendri PAMAM(NHCH2CH2COONa)1024
Molecular Weight:
212790.30
MDL number:
UNSPSC Code:
12162002

description

sodium carboxylate surface groups

Quality Level

form

liquid

mol wt

generation 7.5

feature

core type ethylenediamine core (2-carbon core)

concentration

5 wt. % in methanol

no. Surface Groups

1024

density

0.816 g/mL at 25 °C

storage temp.

2-8°C

Physical form

Contains 1024 surface carboxylate groups.

Legal Information

Manufactured by Dendritech®, Inc.
Dendritech is a registered trademark of Dendritech, Inc.

signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

49.5 °F

flash_point_c

9.7 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service