Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

53676

Sigma-Aldrich

β-Humulene

technical, ≥90% (GC)

Synonym(s):

trans,trans-8-Methylene-1,4,4-trimethyl-1,5-cycloundecadiene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H24
CAS Number:
Molecular Weight:
204.35
Beilstein/REAXYS Number:
3238625
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

grade

technical

assay

≥90% (GC)

refractive index

n20/D 1.504

density

0.89 g/mL at 20 °C (lit.)

storage temp.

2-8°C

SMILES string

C\C1=C/CC(C)(C)\C=C\CC(=C)CCC1

InChI

1S/C15H24/c1-13-7-5-8-14(2)10-12-15(3,4)11-6-9-13/h6,10-11H,1,5,7-9,12H2,2-4H3/b11-6+,14-10+

InChI key

HAVYZKHVTLAPDZ-PPGMXFKZSA-N

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

194.0 °F - closed cup

flash_point_c

90 °C - closed cup

ppe

Eyeshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service