Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

533734

Sigma-Aldrich

1,3-Propanediol

≥99.6%

Synonym(s):

1,3-Dihydroxypropane, Trimethylene glycol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HO(CH2)3OH
CAS Number:
Molecular Weight:
76.09
Beilstein/REAXYS Number:
969155
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:

vapor pressure

0.8 mmHg ( 20 °C)
9.8 mmHg ( 100 °C)

assay

≥99.6%

impurities

<0.1% water

refractive index

n20/D 1.440 (lit.)

surface tension

46.2 dyn/cm, 20 °C

viscosity

52 cP(20 °C)(lit.)

bp

214 °C/760 mmHg (lit.)

mp

−27 °C (lit.)

solubility

H2O: soluble

density

1.053 g/mL at 25 °C (lit.)

anion traces

chloride (Cl-): <0.5 ppm

SMILES string

OCCCO[H]

InChI

1S/C3H8O2/c4-2-1-3-5/h4-5H,1-3H2

InChI key

YPFDHNVEDLHUCE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

For applications, see P5040-4

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Celia L Menckeberg et al.
Gut, 64(6), 884-893 (2014-07-25)
Repetitive interaction with microbial stimuli renders epithelial cells (ECs) hyporesponsive to microbial stimulation. Previously, we have reported that buccal ECs from a subset of paediatric patients with Crohn's disease are not hyporesponsive and spontaneously released chemokines. We now aimed to
Sarbjit S Saini et al.
The Journal of investigative dermatology, 135(1), 67-75 (2014-07-22)
ASTERIA I was a 40-week, randomized, double-blind, placebo-controlled study to evaluate the efficacy and safety of subcutaneous omalizumab as add-on therapy for 24 weeks in patients with chronic idiopathic urticaria/spontaneous urticaria (CIU/CSU) who remained symptomatic despite H1 antihistamine treatment at
E Celińska
Biotechnology advances, 28(4), 519-530 (2010-04-07)
The history of 1,3-propanediol (1,3-PD) conversion from being a specialty chemical to being a bulk chemical illustrates that the concerted effort of different metabolic engineering approaches brings the most successful results. In order to metabolically tailor the 1,3-PD production pathway
R K Saxena et al.
Biotechnology advances, 27(6), 895-913 (2009-08-12)
1,3-Propanediol, a valuable bifunctional molecule, can be produced from renewable resources using microorganisms. It has several promising properties for many synthetic reactions, particularly for polymer and cosmetic industries. By virtue of being a natural product, relevant biochemical pathways can be
Charles E Nakamura et al.
Current opinion in biotechnology, 14(5), 454-459 (2003-10-29)
Improvements in the biological production of 1,3-propanediol, a key component of an emerging polymer business, have been realized. Utilizing genes from natural strains that produce 1,3-propanediol from glycerol, metabolic engineering has enabled the development of a recombinant strain that utilizes

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service