Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

533246

Sigma-Aldrich

2-Fluoro-3-methylpyridine

97%

Synonym(s):

2-Fluoro-3-picoline

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H6FN
CAS Number:
Molecular Weight:
111.12
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

97%

refractive index

n20/D 1.476 (lit.)

bp

154-155 °C (lit.)

density

1.098 g/mL at 25 °C (lit.)

SMILES string

Cc1cccnc1F

InChI

1S/C6H6FN/c1-5-3-2-4-8-6(5)7/h2-4H,1H3

InChI key

PHGXUFPYCAWEHK-UHFFFAOYSA-N

General description

2-Fluoro-3-methylpyridine, also known as 2-fluoro-3-picoline, can be prepared by reacting 2-amino-3-methylpyridine and 40% fluoroboric acid.2 The signs of the long range coupling constants of 2-fluoro-3-methylpyridine have been obtained by double and triple resonance techniques. [1][2]

Application

2-Fluoro-3-methylpyridine may be used in the preparation of:
  • 2-phenyl-3-methylpyridine[3]
  • 2-(4-chlorophenyl)-3-methylpyridine[1]
  • 2-(4-fluorophenyl)-3-methylpyridine[2]
  • 2-fluoronicotinic acid[4]

pictograms

FlameExclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

118.9 °F - closed cup

flash_point_c

48.3 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Proton Magnetic Resonance of the Methyl Derivatives of 2-Fluoropyridine. Sigma and Pi Electron Contributions to Spin-Spin Coupling Constants.
Rowbotham JB, et al.
Canadian Journal of Chemistry, 49(11), 1799-1803 (1971)
Signs of proton-proton and proton-fluorine spin-spin coupling constants in 2-fluoro-3-methylpyridine. Sigma and pi contributions to coupling in a nitrogen heterocycle.
Schaefer T, et al.
Canadian Journal of Chemistry, 47(9), 1507-1514 (1969)
Synthesis of 2-and 6-fluoronicotinamides.
Minor JT, et al.
Journal of the American Chemical Society, 71(3), 1125-1126 (1949)
A facile synthesis of 7-halo-5H-indeno [1, 2-b] pyridines and-pyridin-5-ones.
DuPriest MT, et al.
The Journal of Organic Chemistry, 51(11), 2021-2023 (1986)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service