532509
(S)-(−)-Trifluorolactic acid
98%
Synonym(s):
(S)-(−)-3,3,3-Trifluoro-2-hydroxypropanoic acid
About This Item
Recommended Products
assay
98%
optical activity
[α]20/D −42°, c = 1 in chloroform
mp
72-76 °C (lit.)
SMILES string
O[C@@H](C(O)=O)C(F)(F)F
InChI
1S/C3H3F3O3/c4-3(5,6)1(7)2(8)9/h1,7H,(H,8,9)/t1-/m0/s1
InChI key
BVKGUTLIPHZYCX-SFOWXEAESA-N
Application
It can also be utilized as a reactant to synthesize:
- (S,S) Double-headed bis(trifluorolactate)s with polymethylene chains by esterification reaction with α,ω-alkanediols.[2]
- Chiral 1,1,1-trifluoromethyl-2-alkanols.[1]
signalword
Danger
hcodes
Hazard Classifications
Eye Dam. 1 - Skin Corr. 1B
Storage Class
8A - Combustible corrosive hazardous materials
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Faceshields, Gloves, Goggles, type ABEK (EN14387) respirator filter
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service