Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

530859

Sigma-Aldrich

exo-2-Norbornylisothiocyanate

96%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H11NS
CAS Number:
Molecular Weight:
153.24
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

96%

refractive index

n20/D 1.553 (lit.)

bp

81-83 °C/2 mmHg (lit.)

mp

27-30 °C (lit.)

functional group

isothiocyanate

SMILES string

[H][C@@]12CC[C@@]([H])(C1)[C@H](C2)N=C=S

InChI

1S/C8H11NS/c10-5-9-8-4-6-1-2-7(8)3-6/h6-8H,1-4H2/t6-,7+,8+/m1/s1

InChI key

RBGDFYZLQKZUCO-CSMHCCOUSA-N

General description

exo-2-Norbornylisothiocyanate can be formed by the isomerization of exo-2-norbornylthiocyanate, which proceeds through the formation of non-classical ion.

pictograms

Corrosion

signalword

Danger

hcodes

Hazard Classifications

Skin Corr. 1B

Storage Class

8A - Combustible corrosive hazardous materials

wgk_germany

WGK 3

flash_point_f

226.0 °F - closed cup

flash_point_c

107.8 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type P3 (EN 143) respirator cartridges


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service