Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

529125

Sigma-Aldrich

Methyl 1-methoxy-2-naphthoate

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3OC10H6CO2CH3
CAS Number:
Molecular Weight:
216.23
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

98%

refractive index

n20/D 1.603 (lit.)

bp

320 °C (lit.)

density

1.17 g/mL at 25 °C (lit.)

SMILES string

COC(=O)c1ccc2ccccc2c1OC

InChI

1S/C13H12O3/c1-15-12-10-6-4-3-5-9(10)7-8-11(12)13(14)16-2/h3-8H,1-2H3

InChI key

IKYRLYKWULPUNA-UHFFFAOYSA-N

General description

Methyl 1-methoxy-2-naphthoate is an aromatic ester.

Application

Methyl 1-methoxy-2-naphthoate may be used to synthesize a precursor, required for the preparation of a naphthopyran derivative[1] and (3,4-dihydro-3,3-dimethyl-2H-naphtho[2,3-b]pyran-5,10-dione).[2]

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

O Kodama et al.
Journal of natural products, 56(2), 292-294 (1993-02-01)
Rhinacanthus nasutus (Acanthaceae) gave a new antifungal naphthopyran derivative. The structure of the compound was elucidated by spectroscopic methods as 3,4-dihydro-3,3-dimethyl-2H-naphtho[2,3-b]pyran-5,10-dione [1].
Synthesis of an antifungal naphthopyran derivative isolated from Rhinacanthus nasutus (Acanthaceae).
Kuwahara S, et al.
Agricultural and Biological Chemistry, 55(11), 2909-2911 (1991)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service