528536
Methyl 2-methoxy-1-naphthoate
98%
Synonym(s):
Methyl 2-methoxynaphthoate
About This Item
assay
98%
bp
120-121 °C/0.05 mmHg (lit.)
mp
50-53 °C (lit.)
SMILES string
COC(=O)c1c(OC)ccc2ccccc12
InChI
1S/C13H12O3/c1-15-11-8-7-9-5-3-4-6-10(9)12(11)13(14)16-2/h3-8H,1-2H3
InChI key
BDXGQHHFVPQVAQ-UHFFFAOYSA-N
General description
Application
Storage Class
11 - Combustible Solids
wgk_germany
WGK 3
flash_point_f
230.0 °F - closed cup
flash_point_c
110 °C - closed cup
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service