Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

528064

Sigma-Aldrich

Vinyl chloroformate

99%

Synonym(s):

Vinyl chlorocarbonate, Vinyloxycarbonyl chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH2=CHOCOCl
CAS Number:
Molecular Weight:
106.51
Beilstein/REAXYS Number:
1741679
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:

vapor pressure

0.88 psi ( 20 °C)

assay

99%

contains

100 ppm BHT as inhibitor

refractive index

n20/D 1.41 (lit.)

bp

68-69 °C/735 mmHg (lit.)

density

1.166 g/mL at 20 °C (lit.)

storage temp.

2-8°C

SMILES string

ClC(=O)OC=C

InChI

1S/C3H3ClO2/c1-2-6-3(4)5/h2H,1H2

InChI key

PQLFROTZSIMBKR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1B

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

24.8 °F - closed cup

flash_point_c

-4 °C - closed cup

ppe

Faceshields, Gloves, Goggles, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service