526630
3-Chloro-2-fluoro-5-(trifluoromethyl)benzaldehyde
96%
Synonym(s):
5-Chloro-6,α,α,α-tetrafluoro-m-tolualdehyde
About This Item
assay
96%
refractive index
n20/D 1.476 (lit.)
bp
196 °C (lit.)
density
1.54 g/mL at 25 °C (lit.)
SMILES string
Fc1c(Cl)cc(cc1C=O)C(F)(F)F
InChI
1S/C8H3ClF4O/c9-6-2-5(8(11,12)13)1-4(3-14)7(6)10/h1-3H
InChI key
MSZTVIFIFJCNRQ-UHFFFAOYSA-N
General description
signalword
Warning
hcodes
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
target_organs
Respiratory system
Storage Class
10 - Combustible liquids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type ABEK (EN14387) respirator filter
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service