Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

526045

Sigma-Aldrich

trans-2-(4-Biphenyl)vinylboronic acid

≥95%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5C6H4CH=CHB(OH)2
CAS Number:
Molecular Weight:
224.06
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

assay

≥95%

form

crystals

mp

134-139 °C (lit.)

functional group

phenyl

SMILES string

[H]\C(B(O)O)=C(\[H])c1ccc(cc1)-c2ccccc2

InChI

1S/C14H13BO2/c16-15(17)11-10-12-6-8-14(9-7-12)13-4-2-1-3-5-13/h1-11,16-17H/b11-10+

InChI key

NTRGFVQIGQYKIL-ZHACJKMWSA-N

Application

  • Carbonic anhydrase inhibitor
  • Inhibitor of fatty acid amide hydrolase

Other Notes

Contains varying amounts of anhydride

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service