Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

525065

Sigma-Aldrich

Tris(tetramethylcyclopentadienyl)terbium(III)

99.9%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H39Tb
CAS Number:
Molecular Weight:
522.52
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

assay

99.9%

reaction suitability

core: terbium
reagent type: catalyst

mp

175 °C (lit.)

SMILES string

CC1=C(C)C(C)=C(C)C1[Tb](C2C(C)=C(C)C(C)=C2C)C3C(C)=C(C)C(C)=C3C

InChI

1S/3C9H13.Tb/c3*1-6-5-7(2)9(4)8(6)3;/h3*5H,1-4H3;

Inchi Key

VTIJOFRWKLSSMD-UHFFFAOYSA-N

General description

Atomic number of base material: 65 Terbium

Pictograms

Flame

signalword

Danger

hcodes

pcodes

Hazard Classifications

Flam. Sol. 1

supp_hazards

Storage Class

4.1B - Flammable solid hazardous materials

wgk_germany

WGK 3

flash_point_f

149.4 °F - closed cup

flash_point_c

65.2 °C - closed cup

ppe

Eyeshields, Gloves, type P3 (EN 143) respirator cartridges


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service