Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

524611

Sigma-Aldrich

JandaJel-NH2

100-200 mesh, extent of labeling: 1.0 mmol/g N loading, 2 % cross-linked

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.22

product line

JandaJel

crosslinking

2 % cross-linked

reaction suitability

reaction type: Fmoc solid-phase peptide synthesis

extent of labeling

1.0 mmol/g N loading

particle size

100-200 mesh

functional group

amine

Looking for similar products? Visit Product Comparison Guide

Legal Information

JandaJel is a trademark of Sigma-Aldrich Co. LLC

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cansu Karabiyik et al.
STAR protocols, 2(4), 100926-100926 (2021-11-13)
The regulation of lipid kinases has remained elusive given the difficulties of assessing changes in lipid levels. Here, we describe the isolation of protein and lipid kinases to determine the regulation of lipid kinases in vitro. This can be followed
Cansu Karabiyik et al.
Developmental cell, 56(13), 1961-1975 (2021-06-10)
Autophagy is an essential catabolic process induced to provide cellular energy sources in response to nutrient limitation through the activation of kinases, like AMP-activated protein kinase (AMPK) and ULK1. Although glucose starvation induces autophagy, the exact mechanism underlying this signaling

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service